| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007434.hg.1 |
ENST00000558104.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007434.hg.1 |
ENST00000558104.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007434.hg.1 |
ENST00000558104.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007434.hg.1 |
ENST00000558104.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007434.hg.1 |
ENST00000561413.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007434.hg.1 |
ENST00000561413.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007434.hg.1 |
ENST00000561413.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007434.hg.1 |
NR_120318.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007434.hg.1 |
NR_120318.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007434.hg.1 |
NR_120318.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007434.hg.1 |
NR_120318.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007434.hg.1 |
NR_120318.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007434.hg.1 |
NR_120318.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007434.hg.1 |
NR_120319.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007434.hg.1 |
NR_120319.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007434.hg.1 |
NR_120319.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007434.hg.1 |
NR_120319.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007434.hg.1 |
NR_120319.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500152851.hg.1 |
ENST00000558104.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500152851.hg.1 |
NR_120318.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500152851.hg.1 |
NR_120319.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500152851.hg.1 |
ENST00000558104.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500152851.hg.1 |
NR_120318.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500152851.hg.1 |
NR_120319.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500152851.hg.1 |
TC1500007434.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1500007434.hg.1 |
ENST00000558104.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007434.hg.1 |
ENST00000558104.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007434.hg.1 |
ENST00000558104.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007434.hg.1 |
ENST00000558104.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007434.hg.1 |
ENST00000561413.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007434.hg.1 |
ENST00000561413.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007434.hg.1 |
ENST00000561413.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007434.hg.1 |
NR_120318.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007434.hg.1 |
NR_120318.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007434.hg.1 |
NR_120318.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007434.hg.1 |
NR_120318.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007434.hg.1 |
NR_120318.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007434.hg.1 |
NR_120318.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007434.hg.1 |
NR_120319.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007434.hg.1 |
NR_120319.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007434.hg.1 |
NR_120319.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007434.hg.1 |
NR_120319.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007434.hg.1 |
NR_120319.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500152852.hg.1 |
NR_120318.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500152852.hg.1 |
NR_120319.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500152852.hg.1 |
NR_120318.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500152852.hg.1 |
NR_120319.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500152852.hg.1 |
TC1500007434.hg.1 |
Clariom D TC |