Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
ENST00000005558.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
ENST00000403825.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
ENST00000429071.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
ENST00000440625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
ENST00000466459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
ENST00000476927.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
ENST00000535603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
ENST00000621379.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
NM_001007245.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
NM_001197079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
NM_001197080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
NM_001550.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
NR_120333.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
ENST00000005558.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
ENST00000403825.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
ENST00000429071.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
ENST00000440625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
ENST00000466459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
ENST00000476927.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
ENST00000535603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
ENST00000621379.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
NM_001007245.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
NM_001197079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
NM_001197080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
NM_001550.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
NR_120333.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198575.hg.1 |
TC0700013441.hg.1 |
Clariom D TC |