Assay Details
| Gene Symbol: RORA-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120339.1 | 2 - 3 | 294 | 103 | ||
| NR_120340.1 | 2 - 3 | 294 | 103 | |||
| NR_120342.1 | 2 - 3 | 294 | 103 | |||
| GenBank mRNA | BC035094.2 | - | 2 - 3 | 291 | 103 | |
| BI668481.1 | - | 2 - 3 | 300 | 103 | ||
| BX115309.1 | - | 2 - 3 | 123 | 103 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | interactor of little elongation complex ELL subunit 2 | ICE2 |
BRCC1, NARG2 |
-9.4kb | Anti-sense | Chr. 15 - 60419609 - 60479160 on NCBI Build GRCh38 |
| Downstream | RORA antisense RNA 2 | RORA-AS2 | 171.0kb | Sense | Chr. 15 - 60681569 - 60687249 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

RORA-AS1
Genomic Map