| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000561571.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000561571.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000561571.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000561571.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000561571.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000561571.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000561571.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000561571.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000562634.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000562634.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000562634.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000562634.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000562634.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000564562.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000564562.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000564562.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000566268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000566268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000566268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000566268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000566268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000566268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000566268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000566268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000569258.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000569258.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
ENST00000569258.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
NR_120348.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
NR_120348.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
NR_120348.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
NR_120348.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
NR_120348.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
NR_120348.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
NR_120349.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
NR_120349.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
NR_120349.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
NR_120349.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
NR_120350.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
NR_120350.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
NR_120350.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
NR_120350.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
NR_120350.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
NR_120351.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
NR_120351.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
NR_120351.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
NR_120351.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009933.hg.1 |
NR_120351.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500173664.hg.1 |
ENST00000561571.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500173664.hg.1 |
ENST00000561571.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500173664.hg.1 |
NR_120348.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500173664.hg.1 |
TC1500009933.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500173664.hg.1 |
ENST00000561571.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500173664.hg.1 |
ENST00000561571.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500173664.hg.1 |
NR_120348.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000561571.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000561571.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000561571.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000561571.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000561571.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000561571.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000561571.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000561571.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000562634.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000562634.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000562634.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000562634.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000562634.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000564562.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000564562.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000564562.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000566268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000566268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000566268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000566268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000566268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000566268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000566268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000566268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000569258.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000569258.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
ENST00000569258.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
NR_120348.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
NR_120348.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
NR_120348.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
NR_120348.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
NR_120348.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
NR_120348.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
NR_120349.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
NR_120349.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
NR_120349.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
NR_120349.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
NR_120350.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
NR_120350.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
NR_120350.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
NR_120350.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
NR_120350.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
NR_120351.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
NR_120351.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
NR_120351.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
NR_120351.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009933.hg.1 |
NR_120351.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500173667.hg.1 |
ENST00000561571.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500173667.hg.1 |
ENST00000561571.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500173667.hg.1 |
ENST00000564562.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500173667.hg.1 |
ENST00000566268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500173667.hg.1 |
ENST00000566268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500173667.hg.1 |
NR_120348.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500173667.hg.1 |
NR_120350.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500173667.hg.1 |
NR_120351.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500173667.hg.1 |
TC1500009933.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500173667.hg.1 |
ENST00000561571.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500173667.hg.1 |
ENST00000561571.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500173667.hg.1 |
ENST00000564562.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500173667.hg.1 |
ENST00000566268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500173667.hg.1 |
ENST00000566268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500173667.hg.1 |
NR_120348.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500173667.hg.1 |
NR_120350.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500173667.hg.1 |
NR_120351.1 |
Refseq |