Assay Details
| Gene Symbol: THSD4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120349.1 | 1 - 2 | 243 | 104 | ||
| GenBank mRNA | DA296094.1 | - | 1 - 2 | 179 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cancer/testis antigen 62 | CT62 | -51.3kb | Sense | Chr. 15 - 71110244 - 71115551 on NCBI Build GRCh38 | |
| Downstream | THSD4 antisense RNA 2 | THSD4-AS2 | 499.6kb | Sense | Chr. 15 - 71688701 - 71691539 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000561571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000561571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000561571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000561571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000561571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000561571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000561571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000561571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000562634.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000562634.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000562634.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000562634.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000562634.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000564562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000564562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000564562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000569258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000569258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | ENST00000569258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | NR_120348.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | NR_120348.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | NR_120348.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | NR_120348.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | NR_120348.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | NR_120348.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | NR_120349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | NR_120349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | NR_120349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | NR_120349.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | NR_120350.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | NR_120350.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | NR_120350.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | NR_120350.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | NR_120350.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | NR_120351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | NR_120351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | NR_120351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | NR_120351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009933.hg.1 | NR_120351.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173661.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173661.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173661.hg.1 | ENST00000569258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173661.hg.1 | NR_120348.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173661.hg.1 | NR_120349.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173661.hg.1 | NR_120350.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173661.hg.1 | NR_120351.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173661.hg.1 | TC1500009933.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500173661.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173661.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173661.hg.1 | ENST00000569258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173661.hg.1 | NR_120348.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173661.hg.1 | NR_120349.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173661.hg.1 | NR_120350.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173661.hg.1 | NR_120351.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173659.hg.1 | NR_120348.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173659.hg.1 | NR_120349.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173659.hg.1 | NR_120350.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173659.hg.1 | NR_120351.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173659.hg.1 | TC1500009933.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500173659.hg.1 | NR_120348.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173659.hg.1 | NR_120349.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173659.hg.1 | NR_120350.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173659.hg.1 | NR_120351.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173673.hg.1 | ENST00000561571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173673.hg.1 | ENST00000561571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173673.hg.1 | ENST00000562634.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173673.hg.1 | ENST00000564562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173673.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173673.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173673.hg.1 | ENST00000569258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173673.hg.1 | NR_120348.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173673.hg.1 | NR_120349.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173673.hg.1 | NR_120350.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173673.hg.1 | NR_120351.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173673.hg.1 | TC1500009933.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500173673.hg.1 | ENST00000561571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173673.hg.1 | ENST00000561571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173673.hg.1 | ENST00000562634.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173673.hg.1 | ENST00000564562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173673.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173673.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173673.hg.1 | ENST00000569258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173673.hg.1 | NR_120348.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173673.hg.1 | NR_120349.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173673.hg.1 | NR_120350.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173673.hg.1 | NR_120351.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000561571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000561571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000561571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000561571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000561571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000561571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000561571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000561571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000562634.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000562634.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000562634.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000562634.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000562634.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000564562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000564562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000564562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000569258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000569258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | ENST00000569258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | NR_120348.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | NR_120348.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | NR_120348.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | NR_120348.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | NR_120348.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | NR_120348.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | NR_120349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | NR_120349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | NR_120349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | NR_120349.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | NR_120350.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | NR_120350.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | NR_120350.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | NR_120350.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | NR_120350.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | NR_120351.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | NR_120351.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | NR_120351.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | NR_120351.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009933.hg.1 | NR_120351.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173662.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173662.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173662.hg.1 | ENST00000569258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173662.hg.1 | NR_120348.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173662.hg.1 | NR_120349.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173662.hg.1 | NR_120350.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173662.hg.1 | NR_120351.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173662.hg.1 | TC1500009933.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500173662.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173662.hg.1 | ENST00000566268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173662.hg.1 | ENST00000569258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173662.hg.1 | NR_120348.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173662.hg.1 | NR_120349.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173662.hg.1 | NR_120350.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173662.hg.1 | NR_120351.1 | Refseq |

THSD4-AS1
Genomic Map