| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000559277.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000559277.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000560851.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000560851.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000560851.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000560851.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000560851.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000560851.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000560851.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000560973.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000560973.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
ENST00000560973.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
NR_120365.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
NR_120365.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
NR_120365.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
NR_120365.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
NR_120365.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
NR_120365.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
NR_120365.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
NR_120365.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
NR_120365.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
NR_120365.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008062.hg.1 |
NR_120365.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000559277.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000559277.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000560851.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000560851.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000560851.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000560851.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000560851.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000560851.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000560851.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000560973.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000560973.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
ENST00000560973.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
NR_120365.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
NR_120365.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
NR_120365.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
NR_120365.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
NR_120365.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
NR_120365.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
NR_120365.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
NR_120365.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
NR_120365.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
NR_120365.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008062.hg.1 |
NR_120365.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500158001.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500158001.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500158001.hg.1 |
ENST00000560851.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500158001.hg.1 |
NR_120365.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500158001.hg.1 |
TC1500008062.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500158001.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500158001.hg.1 |
ENST00000559781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500158001.hg.1 |
ENST00000560851.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500158001.hg.1 |
NR_120365.1 |
Refseq |