| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010443.hg.1 |
ENST00000558105.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010443.hg.1 |
ENST00000558105.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010443.hg.1 |
ENST00000558105.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010443.hg.1 |
ENST00000558105.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010443.hg.1 |
ENST00000559531.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010443.hg.1 |
ENST00000559531.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010443.hg.1 |
ENST00000559531.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010443.hg.1 |
ENST00000559531.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010443.hg.1 |
ENST00000559531.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010443.hg.1 |
NR_120372.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010443.hg.1 |
NR_120372.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010443.hg.1 |
NR_120372.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010443.hg.1 |
NR_120372.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010443.hg.1 |
NR_120372.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500178662.hg.1 |
ENST00000559531.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500178662.hg.1 |
NR_120372.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500178662.hg.1 |
TC1500010443.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500178662.hg.1 |
ENST00000559531.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500178662.hg.1 |
NR_120372.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010443.hg.1 |
ENST00000558105.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010443.hg.1 |
ENST00000558105.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010443.hg.1 |
ENST00000558105.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010443.hg.1 |
ENST00000558105.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010443.hg.1 |
ENST00000559531.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010443.hg.1 |
ENST00000559531.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010443.hg.1 |
ENST00000559531.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010443.hg.1 |
ENST00000559531.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010443.hg.1 |
ENST00000559531.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010443.hg.1 |
NR_120372.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010443.hg.1 |
NR_120372.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010443.hg.1 |
NR_120372.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010443.hg.1 |
NR_120372.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010443.hg.1 |
NR_120372.1 |
Refseq |