| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008840.hg.1 |
ENST00000333666.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008840.hg.1 |
ENST00000333666.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008840.hg.1 |
ENST00000333666.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008840.hg.1 |
ENST00000333666.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008840.hg.1 |
ENST00000333666.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008840.hg.1 |
ENST00000333666.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008840.hg.1 |
ENST00000333666.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008840.hg.1 |
ENST00000567588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008840.hg.1 |
ENST00000567588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008840.hg.1 |
ENST00000567588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008840.hg.1 |
ENST00000567588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008840.hg.1 |
ENST00000567588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008840.hg.1 |
ENST00000567588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008840.hg.1 |
NR_120387.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008840.hg.1 |
NR_120387.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008840.hg.1 |
NR_120387.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008840.hg.1 |
NR_120387.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008840.hg.1 |
NR_120387.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008840.hg.1 |
NR_120387.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008840.hg.1 |
NR_120387.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600165873.hg.1 |
ENST00000333666.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600165873.hg.1 |
NR_120387.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600165873.hg.1 |
TC1600008840.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600165873.hg.1 |
ENST00000333666.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600165873.hg.1 |
NR_120387.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600165878.hg.1 |
ENST00000333666.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600165878.hg.1 |
ENST00000567588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600165878.hg.1 |
NR_120387.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600165878.hg.1 |
TC1600008840.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600165878.hg.1 |
ENST00000333666.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600165878.hg.1 |
ENST00000567588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600165878.hg.1 |
NR_120387.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600165879.hg.1 |
ENST00000333666.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600165879.hg.1 |
ENST00000567588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600165879.hg.1 |
NR_120387.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600165879.hg.1 |
TC1600008840.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600165879.hg.1 |
ENST00000333666.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600165879.hg.1 |
ENST00000567588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600165879.hg.1 |
NR_120387.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008840.hg.1 |
ENST00000333666.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008840.hg.1 |
ENST00000333666.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008840.hg.1 |
ENST00000333666.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008840.hg.1 |
ENST00000333666.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008840.hg.1 |
ENST00000333666.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008840.hg.1 |
ENST00000333666.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008840.hg.1 |
ENST00000333666.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008840.hg.1 |
ENST00000567588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008840.hg.1 |
ENST00000567588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008840.hg.1 |
ENST00000567588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008840.hg.1 |
ENST00000567588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008840.hg.1 |
ENST00000567588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008840.hg.1 |
ENST00000567588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008840.hg.1 |
NR_120387.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008840.hg.1 |
NR_120387.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008840.hg.1 |
NR_120387.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008840.hg.1 |
NR_120387.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008840.hg.1 |
NR_120387.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008840.hg.1 |
NR_120387.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008840.hg.1 |
NR_120387.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600165875.hg.1 |
ENST00000333666.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600165875.hg.1 |
ENST00000567588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600165875.hg.1 |
NR_120387.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600165875.hg.1 |
TC1600008840.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600165875.hg.1 |
ENST00000333666.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600165875.hg.1 |
ENST00000567588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600165875.hg.1 |
NR_120387.1 |
Refseq |