Assay Details
| Gene Symbol: NUBPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120408.1 | 6 - 7 | 565 | 105 | ||
| GenBank mRNA | BX248028.1 | - | 6 - 7 | 552 | 105 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | G protein-coupled receptor 33 (gene/pseudogene) | GPR33 | -185.5kb | Anti-sense | Chr. 14 - 31482944 - 31487945 on NCBI Build GRCh38 | |
| Downstream | ARHGAP5 antisense RNA 1 (head to head) | ARHGAP5-AS1 | C14orf128 | 248.7kb | Anti-sense | Chr. 14 - 32075419 - 32076699 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

NUBPL
Genomic Map