| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006636.hg.1 |
ENST00000457628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006636.hg.1 |
ENST00000457628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006636.hg.1 |
ENST00000457628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006636.hg.1 |
ENST00000457628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006636.hg.1 |
NR_120427.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006636.hg.1 |
NR_120427.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006636.hg.1 |
NR_120427.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006636.hg.1 |
NR_120427.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006636.hg.1 |
NR_120427.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006636.hg.1 |
NR_120427.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008383.hg.1 |
ENST00000623176.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300145279.hg.1 |
NR_120427.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300145279.hg.1 |
NR_120427.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300145279.hg.1 |
TC1300006636.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300145278.hg.1 |
ENST00000457628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300145278.hg.1 |
NR_120427.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300145278.hg.1 |
ENST00000457628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300145278.hg.1 |
NR_120427.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300145278.hg.1 |
TC1300006636.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300008383.hg.1 |
ENST00000623176.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006636.hg.1 |
ENST00000457628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006636.hg.1 |
ENST00000457628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006636.hg.1 |
ENST00000457628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006636.hg.1 |
ENST00000457628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006636.hg.1 |
NR_120427.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006636.hg.1 |
NR_120427.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006636.hg.1 |
NR_120427.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006636.hg.1 |
NR_120427.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006636.hg.1 |
NR_120427.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006636.hg.1 |
NR_120427.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300145277.hg.1 |
ENST00000457628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300145277.hg.1 |
NR_120427.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300145277.hg.1 |
ENST00000457628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300145277.hg.1 |
NR_120427.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300145277.hg.1 |
TC1300006636.hg.1 |
Clariom D TC |