| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008382.hg.1 |
ENST00000413501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008382.hg.1 |
ENST00000413501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008382.hg.1 |
ENST00000413501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008382.hg.1 |
ENST00000413501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008382.hg.1 |
ENST00000413501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008382.hg.1 |
ENST00000413501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008382.hg.1 |
NR_120428.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008382.hg.1 |
NR_120428.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008382.hg.1 |
NR_120428.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008382.hg.1 |
NR_120428.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008382.hg.1 |
NR_120428.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156588.hg.1 |
ENST00000413501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300156588.hg.1 |
NR_120428.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156588.hg.1 |
ENST00000413501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300156588.hg.1 |
NR_120428.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156588.hg.1 |
TC1300008382.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300008382.hg.1 |
ENST00000413501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008382.hg.1 |
ENST00000413501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008382.hg.1 |
ENST00000413501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008382.hg.1 |
ENST00000413501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008382.hg.1 |
ENST00000413501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008382.hg.1 |
ENST00000413501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008382.hg.1 |
NR_120428.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008382.hg.1 |
NR_120428.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008382.hg.1 |
NR_120428.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008382.hg.1 |
NR_120428.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008382.hg.1 |
NR_120428.1 |
Refseq |