| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012901.hg.1 |
ENST00000526254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012901.hg.1 |
ENST00000526254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012901.hg.1 |
ENST00000526254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012901.hg.1 |
NR_120444.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012901.hg.1 |
NR_120444.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012901.hg.1 |
NR_120444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100198828.hg.1 |
ENST00000526254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100198828.hg.1 |
NR_120444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100198828.hg.1 |
ENST00000526254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100198828.hg.1 |
NR_120444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100198828.hg.1 |
TC1100012901.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100198823.hg.1 |
ENST00000526254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100198823.hg.1 |
NR_120444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100198823.hg.1 |
ENST00000526254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100198823.hg.1 |
NR_120444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100198823.hg.1 |
TC1100012901.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100198831.hg.1 |
ENST00000526254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100198831.hg.1 |
NR_120444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100198831.hg.1 |
ENST00000526254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100198831.hg.1 |
NR_120444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100198831.hg.1 |
TC1100012901.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100012901.hg.1 |
ENST00000526254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012901.hg.1 |
ENST00000526254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012901.hg.1 |
ENST00000526254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012901.hg.1 |
NR_120444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012901.hg.1 |
NR_120444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012901.hg.1 |
NR_120444.1 |
Refseq |