| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009135.hg.1 |
ENST00000437593.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009135.hg.1 |
ENST00000437593.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009135.hg.1 |
NR_120495.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009135.hg.1 |
NR_120495.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009135.hg.1 |
ENST00000437593.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009135.hg.1 |
ENST00000437593.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009135.hg.1 |
NR_120495.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009135.hg.1 |
NR_120495.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000162520.hg.1 |
ENST00000437593.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000162520.hg.1 |
NR_120495.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000162520.hg.1 |
ENST00000437593.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000162520.hg.1 |
NR_120495.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000162520.hg.1 |
TC1000009135.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000162526.hg.1 |
ENST00000437593.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000162526.hg.1 |
NR_120495.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000162526.hg.1 |
ENST00000437593.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000162526.hg.1 |
NR_120495.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000162526.hg.1 |
TC1000009135.hg.1 |
Clariom D TC |