| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000517732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000517732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000517732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000517732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000517732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000517732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000517732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000517732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000517732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000517732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000518098.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000518098.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000518098.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
NR_120604.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
NR_120604.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
NR_120604.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
NR_120604.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
NR_120604.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
NR_120604.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012262.hg.1 |
NR_120604.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800184540.hg.1 |
ENST00000517732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800184540.hg.1 |
ENST00000517732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800184540.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800184540.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800184540.hg.1 |
NR_120604.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800184540.hg.1 |
ENST00000517732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800184540.hg.1 |
ENST00000517732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800184540.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800184540.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800184540.hg.1 |
NR_120604.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800184540.hg.1 |
TC0800012262.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000517732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000517732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000517732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000517732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000517732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000517732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000517732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000517732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000517732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000517732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000518098.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000518098.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000518098.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
ENST00000524047.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
NR_120604.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
NR_120604.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
NR_120604.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
NR_120604.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
NR_120604.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
NR_120604.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012262.hg.1 |
NR_120604.1 |
Refseq |