Assay Details
| Gene Symbol: LINCR-0001 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120604.1 | 4 - 5 | 432 | 125 | ||
| GenBank mRNA | DA212384.1 | - | 4 - 5 | 432 | 125 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | methionine sulfoxide reductase A | MSRA | PMSR | -51.2kb | Sense | Chr. 8 - 10054224 - 10428891 on NCBI Build GRCh38 |
| Downstream |
protease, serine 55 |
PRSS55 |
CT153, T-SP1, TSP1, UNQ9391 |
43.5kb | Sense | Chr. 8 - 10525331 - 10554166 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000518098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000518098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000518098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | NR_120604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | NR_120604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | NR_120604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | NR_120604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | NR_120604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | NR_120604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012262.hg.1 | NR_120604.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184542.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184542.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184542.hg.1 | ENST00000518098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184542.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184542.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184542.hg.1 | NR_120604.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184542.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184542.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184542.hg.1 | ENST00000518098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184542.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184542.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184542.hg.1 | NR_120604.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184542.hg.1 | TC0800012262.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800184545.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184545.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184545.hg.1 | NR_120604.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184545.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184545.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184545.hg.1 | NR_120604.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184545.hg.1 | TC0800012262.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800184541.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184541.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184541.hg.1 | ENST00000518098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184541.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184541.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184541.hg.1 | NR_120604.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184541.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184541.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184541.hg.1 | ENST00000518098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184541.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184541.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184541.hg.1 | NR_120604.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184541.hg.1 | TC0800012262.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000517732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000518098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000518098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000518098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | ENST00000524047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | NR_120604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | NR_120604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | NR_120604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | NR_120604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | NR_120604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | NR_120604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012262.hg.1 | NR_120604.1 | Refseq |

LINCR-0001
Genomic Map