| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000517925.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000517925.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000519062.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000519062.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000519062.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000519062.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000519062.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000519062.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000519062.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000519062.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000524286.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000524286.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000524286.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
ENST00000524286.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
NR_039986.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
NR_039986.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
NR_039986.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
NR_039986.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
NR_039986.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
NR_039986.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
NR_039986.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
NR_039986.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010725.hg.1 |
NR_039986.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
ENST00000438488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
ENST00000438488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
ENST00000438488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
ENST00000438488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
ENST00000438488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
ENST00000439804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
ENST00000439804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
ENST00000439804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
ENST00000439804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
ENST00000439804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
ENST00000439804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
ENST00000439804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
ENST00000439804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
ENST00000439804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
ENST00000439804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
ENST00000439804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
ENST00000439804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
ENST00000439804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
ENST00000439804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
ENST00000439804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
ENST00000439804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
NR_120606.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
NR_120606.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
NR_120606.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
NR_120606.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
NR_120606.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
NR_120606.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
NR_120606.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
NR_120606.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
NR_120606.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
NR_120606.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
NR_120606.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009868.hg.1 |
NR_120606.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000517925.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000517925.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000518540.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000519062.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000519062.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000519062.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000519062.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000519062.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000519062.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000519062.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000519062.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000524286.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000524286.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000524286.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
ENST00000524286.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
NR_039986.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
NR_039986.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
NR_039986.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
NR_039986.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
NR_039986.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
NR_039986.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
NR_039986.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
NR_039986.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010725.hg.1 |
NR_039986.1 |
Refseq |