| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007476.hg.1 |
ENST00000509844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007476.hg.1 |
ENST00000509844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007476.hg.1 |
ENST00000513392.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007476.hg.1 |
ENST00000513392.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007476.hg.1 |
ENST00000513392.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007476.hg.1 |
ENST00000513392.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007476.hg.1 |
ENST00000513392.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007476.hg.1 |
NR_120607.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007476.hg.1 |
NR_120607.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007476.hg.1 |
NR_120607.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007476.hg.1 |
NR_120607.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007476.hg.1 |
NR_120607.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007476.hg.1 |
NR_120607.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007476.hg.1 |
NR_120607.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007476.hg.1 |
NR_120607.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007476.hg.1 |
NR_120607.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007476.hg.1 |
ENST00000509844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007476.hg.1 |
ENST00000509844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007476.hg.1 |
ENST00000513392.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007476.hg.1 |
ENST00000513392.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007476.hg.1 |
ENST00000513392.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007476.hg.1 |
ENST00000513392.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007476.hg.1 |
ENST00000513392.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007476.hg.1 |
NR_120607.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007476.hg.1 |
NR_120607.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007476.hg.1 |
NR_120607.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007476.hg.1 |
NR_120607.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007476.hg.1 |
NR_120607.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007476.hg.1 |
NR_120607.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007476.hg.1 |
NR_120607.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007476.hg.1 |
NR_120607.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007476.hg.1 |
NR_120607.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500150497.hg.1 |
ENST00000509844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500150497.hg.1 |
NR_120607.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500150497.hg.1 |
ENST00000509844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500150497.hg.1 |
NR_120607.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500150497.hg.1 |
TC0500007476.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500150502.hg.1 |
ENST00000509844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500150502.hg.1 |
NR_120607.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500150502.hg.1 |
ENST00000509844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500150502.hg.1 |
NR_120607.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500150502.hg.1 |
TC0500007476.hg.1 |
Clariom D TC |