Assay Details
| Gene Symbol: PLA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120610.1 | 9 - 10 | 1195 | 99 | ||
| GenBank mRNA | AF035269.1 | - | 9 - 10 | 1160 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ADP-ribosylarginine hydrolase | ADPRH | ARH1 | -35.2kb | Sense | Chr. 3 - 119579433 - 119589954 on NCBI Build GRCh38 |
| Downstream | popeye domain containing 2 | POPDC2 | POP2 | 13.2kb | Anti-sense | Chr. 3 - 119642052 - 119660590 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

PLA1A
Genomic Map