Assay Details
| Gene Symbol: LOC101927523 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120624.1 | 1 - 2 | 143 | 113 | ||
| GenBank mRNA | BC039476.1 | - | 1 - 2 | 140 | 110 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cilia and flagella associated protein 58 | CFAP58 |
C10orf80, CCDC147, bA127L20.4, bA127L20.5, bA554P13.1 |
-21.8kb | Anti-sense | Chr. 10 - 104339124 - 104455090 on NCBI Build GRCh38 |
| Downstream | sortilin related VPS10 domain containing receptor 3 | SORCS3 | SORCS | 160.6kb | Anti-sense | Chr. 10 - 104641101 - 105265242 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101927523
Genomic Map