Assay Details
| Gene Symbol: LOC101928834 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120646.1 | 4 - 5 | 444 | 110 | ||
| GenBank mRNA | BC014575.1 | - | 4 - 5 | 448 | 110 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ADP ribosylation factor like GTPase 5B | ARL5B | ARL8 | -1.0mb | Anti-sense | Chr. 10 - 18659335 - 18681639 on NCBI Build GRCh38 |
| Downstream | plexin domain containing 2 | PLXDC2 | TEM7R | 99.3kb | Anti-sense | Chr. 10 - 19816215 - 20289856 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009967.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009967.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168094.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168094.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168094.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168094.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168094.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168094.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168094.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168094.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168094.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168094.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168094.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168094.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168094.hg.1 | TC1000009967.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000168088.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168088.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168088.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168088.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168088.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168088.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168088.hg.1 | TC1000009967.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000168089.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168089.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168089.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168089.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168089.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168089.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168089.hg.1 | TC1000009967.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000168097.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168097.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168097.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168097.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168097.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168097.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168097.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168097.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168097.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168097.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168097.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168097.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168097.hg.1 | TC1000009967.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000168093.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168093.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168093.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168093.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168093.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168093.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168093.hg.1 | ENST00000423551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168093.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168093.hg.1 | ENST00000431157.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168093.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168093.hg.1 | ENST00000451713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168093.hg.1 | NR_120646.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168093.hg.1 | TC1000009967.hg.1 | Clariom D TC |

LOC101928834
Genomic Map