Assay Details
| Gene Symbol: ZNF503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120651.1 | 2 - 3 | 899 | 72 | ||
| GenBank mRNA | AI522172.1 | - | 2 - 3 | 72 | ||
| BX105802.1 | - | 2 - 3 | 251 | 72 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | catechol-O-methyltransferase domain containing 1 | COMTD1 | -44.0kb | Sense | Chr. 10 - 75233971 - 75236012 on NCBI Build GRCh38 | |
| Downstream | ZNF503 antisense RNA 1 | ZNF503-AS1 | 13.0kb | Anti-sense | Chr. 10 - 75296383 - 75361678 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of cell proliferation
negative regulation of gene expression
neural precursor cell proliferation
G1 to G0 transition involved in cell differentiation
nucleic acid binding
metal ion binding
transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of cell proliferation
negative regulation of gene expression
neural precursor cell proliferation
G1 to G0 transition involved in cell differentiation
nucleic acid binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000445450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000445450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000445450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000445450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000535216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000535216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000535216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000535216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000535216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000535216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000535216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | ENST00000535216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | NR_120651.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | NR_120651.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | NR_120651.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | NR_120651.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | NR_120651.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | NR_120651.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | NR_120651.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | NR_120651.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | NR_120651.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011081.hg.1 | NR_120651.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175587.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175587.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175587.hg.1 | ENST00000445450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175587.hg.1 | NR_120651.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175587.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175587.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175587.hg.1 | ENST00000445450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175587.hg.1 | NR_120651.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175587.hg.1 | TC1000011081.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000175588.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175588.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175588.hg.1 | ENST00000445450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175588.hg.1 | NR_120651.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175588.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175588.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175588.hg.1 | ENST00000445450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175588.hg.1 | NR_120651.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175588.hg.1 | TC1000011081.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000372524.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000418818.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000445450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000445450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000445450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000445450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000535216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000535216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000535216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000535216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000535216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000535216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000535216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | ENST00000535216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | NM_032772.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | NR_120651.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | NR_120651.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | NR_120651.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | NR_120651.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | NR_120651.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | NR_120651.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | NR_120651.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | NR_120651.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | NR_120651.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011081.hg.1 | NR_120651.1 | Refseq |

ZNF503
Genomic Map