Assay Details
| Gene Symbol: TMEM246-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_121575.1 | 1 - 2 | 297 | 99 | ||
| GenBank mRNA | AW451983.1 | - | 1 - 2 | 99 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 246 | TMEM246 | C9orf125 | Anti-sense | Chr. 9 - 101473158 - 101487221 on NCBI Build GRCh38 | |
| Downstream | ring finger protein 20 | RNF20 |
BRE1, BRE1A, hBRE1 |
52.3kb | Sense | Chr. 9 - 101533849 - 101563344 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000424154.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000424154.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000424154.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000424154.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000424154.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000424154.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000425734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000425734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000425734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000425734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000450109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000450109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000450109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000450109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000450109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000450109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000450109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | ENST00000450109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | NR_121573.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | NR_121573.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | NR_121573.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | NR_121573.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | NR_121574.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | NR_121574.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | NR_121574.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | NR_121574.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | NR_121574.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | NR_121574.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | NR_121575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | NR_121575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | NR_121575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | NR_121575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | NR_121575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | NR_121576.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | NR_121576.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008255.hg.1 | NR_121576.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900155310.hg.1 | ENST00000425734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900155310.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900155310.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900155310.hg.1 | NR_121574.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900155310.hg.1 | NR_121575.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900155310.hg.1 | ENST00000425734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900155310.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900155310.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900155310.hg.1 | NR_121574.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900155310.hg.1 | NR_121575.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900155310.hg.1 | TC0900008255.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900155306.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900155306.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900155306.hg.1 | NR_121574.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900155306.hg.1 | NR_121575.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900155306.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900155306.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900155306.hg.1 | NR_121574.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900155306.hg.1 | NR_121575.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900155306.hg.1 | TC0900008255.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900155308.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900155308.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900155308.hg.1 | NR_121574.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900155308.hg.1 | NR_121575.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900155308.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900155308.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900155308.hg.1 | NR_121574.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900155308.hg.1 | NR_121575.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900155308.hg.1 | TC0900008255.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900155309.hg.1 | ENST00000425734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900155309.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900155309.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900155309.hg.1 | NR_121574.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900155309.hg.1 | NR_121575.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900155309.hg.1 | ENST00000425734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900155309.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900155309.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900155309.hg.1 | NR_121574.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900155309.hg.1 | NR_121575.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900155309.hg.1 | TC0900008255.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000424154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000424154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000424154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000424154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000424154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000424154.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000425734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000425734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000425734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000425734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000431507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000450109.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000450109.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000450109.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000450109.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000450109.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000450109.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000450109.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | ENST00000450109.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | NR_121573.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | NR_121573.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | NR_121573.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | NR_121573.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | NR_121574.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | NR_121574.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | NR_121574.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | NR_121574.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | NR_121574.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | NR_121574.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | NR_121575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | NR_121575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | NR_121575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | NR_121575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | NR_121575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | NR_121576.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | NR_121576.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008255.hg.1 | NR_121576.1 | Refseq |

TMEM246-AS1
Genomic Map