| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011939.hg.1 |
ENST00000415062.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011939.hg.1 |
ENST00000415062.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011939.hg.1 |
ENST00000415062.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011939.hg.1 |
NR_121579.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011939.hg.1 |
NR_121579.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011939.hg.1 |
NR_121579.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011939.hg.1 |
ENST00000415062.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011939.hg.1 |
ENST00000415062.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011939.hg.1 |
ENST00000415062.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011939.hg.1 |
NR_121579.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011939.hg.1 |
NR_121579.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011939.hg.1 |
NR_121579.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900181617.hg.1 |
ENST00000415062.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900181617.hg.1 |
NR_121579.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900181617.hg.1 |
ENST00000415062.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900181617.hg.1 |
NR_121579.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900181617.hg.1 |
TC0900011939.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900181619.hg.1 |
ENST00000415062.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900181619.hg.1 |
NR_121579.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900181619.hg.1 |
ENST00000415062.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900181619.hg.1 |
NR_121579.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900181619.hg.1 |
TC0900011939.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900181618.hg.1 |
ENST00000415062.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900181618.hg.1 |
NR_121579.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900181618.hg.1 |
ENST00000415062.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900181618.hg.1 |
NR_121579.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900181618.hg.1 |
TC0900011939.hg.1 |
Clariom D TC |