Assay Details
| Gene Symbol: LOC100507406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_121619.1 | 1 - 2 | 330 | 80 | ||
| GenBank mRNA | BF970945.1 | - | 1 - 2 | 331 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102723649 | LOC102723649 | -55.6kb | Anti-sense | Chr. 6 - 137665646 - 137674554 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC100130476 | LOC100130476 | 84.6kb | Sense | Chr. 6 - 137823669 - 137868233 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | ENST00000419220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | ENST00000419220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | ENST00000419220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | ENST00000419220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | ENST00000419220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | ENST00000419220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | NR_121618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | NR_121618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | NR_121618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | NR_121618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | NR_121618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | NR_121618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | NR_121618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | NR_121618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | NR_121619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | NR_121619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | NR_121619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | NR_121619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | NR_121619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | NR_121619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013313.hg.1 | NR_121619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | ENST00000419220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | ENST00000419220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | ENST00000419220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | ENST00000419220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | ENST00000419220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | ENST00000419220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | NR_121618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | NR_121618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | NR_121618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | NR_121618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | NR_121618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | NR_121618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | NR_121618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | NR_121618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | NR_121619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | NR_121619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | NR_121619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | NR_121619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | NR_121619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | NR_121619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013313.hg.1 | NR_121619.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192123.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192123.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192123.hg.1 | ENST00000419220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192123.hg.1 | NR_121618.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192123.hg.1 | NR_121619.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192123.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192123.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192123.hg.1 | ENST00000419220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192123.hg.1 | NR_121618.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192123.hg.1 | NR_121619.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192123.hg.1 | TC0600013313.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600192122.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192122.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192122.hg.1 | ENST00000419220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192122.hg.1 | NR_121618.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192122.hg.1 | NR_121619.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192122.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192122.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192122.hg.1 | ENST00000419220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192122.hg.1 | NR_121618.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192122.hg.1 | NR_121619.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192122.hg.1 | TC0600013313.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600192119.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192119.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192119.hg.1 | ENST00000419220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192119.hg.1 | NR_121618.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192119.hg.1 | NR_121619.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192119.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192119.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192119.hg.1 | ENST00000419220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192119.hg.1 | NR_121618.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192119.hg.1 | NR_121619.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192119.hg.1 | TC0600013313.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600192120.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192120.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192120.hg.1 | ENST00000419220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192120.hg.1 | NR_121618.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192120.hg.1 | NR_121619.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192120.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192120.hg.1 | ENST00000411615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192120.hg.1 | ENST00000419220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192120.hg.1 | NR_121618.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192120.hg.1 | NR_121619.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192120.hg.1 | TC0600013313.hg.1 | Clariom D TC |

LOC100507406
Genomic Map