Assay Details
| Gene Symbol: LOC101927854 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_121667.1 | 3 - 4 | 261 | 103 | ||
| GenBank mRNA | CB994391.1 | - | 3 - 4 | 287 | 103 | |
| KJ999766.1 | - | 2 - 3 | 185 | 103 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 691 | LINC00691 | -74.7kb | Anti-sense | Chr. 3 - 24099974 - 24103247 on NCBI Build GRCh38 | |
| Downstream | THRB antisense RNA 1 | THRB-AS1 | 304.1kb | Sense | Chr. 3 - 24494087 - 24500011 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101927854
Genomic Map