Assay Details
| Gene Symbol: LOC255187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_122044.1 | 1 - 2 | 355 | 79 | ||
| GenBank mRNA | BX106527.1 | - | 1 - 2 | 217 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SH3 domain and tetratricopeptide repeats 2 | SH3TC2 |
CMT4C, MNMN |
-143.0 | Anti-sense | Chr. 5 - 148982150 - 149063174 on NCBI Build GRCh38 |
| Downstream | actin binding LIM protein family member 3 | ABLIM3 | HMFN1661 | 77.1kb | Sense | Chr. 5 - 149141483 - 149260439 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000507318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000507318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000507318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000507318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000507318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000507318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000507373.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000507373.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000509139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000509139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000509139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000509139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000509139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000512007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000512007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000515304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000515304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000515304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | NR_122044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | NR_122044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | NR_122044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | NR_122044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | NR_122044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009053.hg.1 | NR_122044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163052.hg.1 | ENST00000509139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163052.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163052.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163052.hg.1 | NR_122044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163052.hg.1 | ENST00000509139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163052.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163052.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163052.hg.1 | NR_122044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163052.hg.1 | TC0500009053.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500163055.hg.1 | ENST00000507318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163055.hg.1 | ENST00000507373.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163055.hg.1 | ENST00000509139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163055.hg.1 | ENST00000515304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163055.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163055.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163055.hg.1 | NR_122044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163055.hg.1 | ENST00000507318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163055.hg.1 | ENST00000507373.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163055.hg.1 | ENST00000509139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163055.hg.1 | ENST00000515304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163055.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163055.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163055.hg.1 | NR_122044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163055.hg.1 | TC0500009053.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500163051.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163051.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163051.hg.1 | NR_122044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163051.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163051.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163051.hg.1 | NR_122044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163051.hg.1 | TC0500009053.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000507318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000507318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000507318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000507318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000507318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000507318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000507373.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000507373.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000509139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000509139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000509139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000509139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000509139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000512007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000512007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000515304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000515304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000515304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | NR_122044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | NR_122044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | NR_122044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | NR_122044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | NR_122044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009053.hg.1 | NR_122044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163060.hg.1 | ENST00000507318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163060.hg.1 | ENST00000509139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163060.hg.1 | ENST00000512007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163060.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163060.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163060.hg.1 | NR_122044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163060.hg.1 | ENST00000507318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163060.hg.1 | ENST00000509139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163060.hg.1 | ENST00000512007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163060.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163060.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163060.hg.1 | NR_122044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163060.hg.1 | TC0500009053.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500163054.hg.1 | ENST00000507318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163054.hg.1 | ENST00000509139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163054.hg.1 | ENST00000515304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163054.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163054.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163054.hg.1 | NR_122044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163054.hg.1 | ENST00000507318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163054.hg.1 | ENST00000509139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163054.hg.1 | ENST00000515304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163054.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163054.hg.1 | ENST00000515519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163054.hg.1 | NR_122044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163054.hg.1 | TC0500009053.hg.1 | Clariom D TC |

LOC255187
Genomic Map