| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007508.hg.1 |
ENST00000503051.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007508.hg.1 |
ENST00000503051.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007508.hg.1 |
ENST00000503051.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007508.hg.1 |
ENST00000503051.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007508.hg.1 |
NR_125363.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007508.hg.1 |
NR_125363.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007508.hg.1 |
NR_125363.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151303.hg.1 |
ENST00000503051.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151303.hg.1 |
NR_125363.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151303.hg.1 |
ENST00000503051.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151303.hg.1 |
NR_125363.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151303.hg.1 |
TC0400007508.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400151301.hg.1 |
ENST00000503051.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151301.hg.1 |
NR_125363.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151301.hg.1 |
ENST00000503051.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151301.hg.1 |
NR_125363.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151301.hg.1 |
TC0400007508.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400007508.hg.1 |
ENST00000503051.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007508.hg.1 |
ENST00000503051.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007508.hg.1 |
ENST00000503051.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007508.hg.1 |
ENST00000503051.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007508.hg.1 |
NR_125363.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007508.hg.1 |
NR_125363.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007508.hg.1 |
NR_125363.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151302.hg.1 |
ENST00000503051.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151302.hg.1 |
NR_125363.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151302.hg.1 |
ENST00000503051.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151302.hg.1 |
NR_125363.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151302.hg.1 |
TC0400007508.hg.1 |
Clariom D TC |