Assay Details
| Gene Symbol: MKLN1-AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125364.1 | 5 - 6 | 1008 | 162 | ||
| GenBank mRNA | AK054623.1 | - | 5 - 6 | 1005 | 162 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
long intergenic non-protein coding RNA, p53 induced transcript |
LINC-PINT |
LincRNA-Pint, MKLN1-AS1, PINT |
-199.8kb | Sense | Chr. 7 - 130877562 - 131109916 on NCBI Build GRCh38 |
| Downstream | podocalyxin like | PODXL |
Gp200, PC, PCLP, PCLP-1 |
189.0kb | Sense | Chr. 7 - 131500262 - 131556617 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000416220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000416220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000416220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000416220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000416220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000416220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000416220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000416220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000416220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000416220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000416220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000416220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000447514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000447514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000447514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000447514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000447514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | NR_125364.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | NR_125364.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | NR_125364.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | NR_125364.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | NR_125364.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | NR_125364.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | NR_125364.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | NR_125364.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | NR_125364.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | NR_125364.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | NR_125364.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012633.hg.1 | NR_125364.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700191044.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700191044.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700191044.hg.1 | NR_125364.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700191044.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700191044.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700191044.hg.1 | NR_125364.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700191044.hg.1 | TC0700012633.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000416220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000416220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000416220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000416220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000416220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000416220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000416220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000416220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000416220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000416220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000416220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000416220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000429067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000444245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000447514.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000447514.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000447514.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000447514.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000447514.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000447904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | ENST00000454515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | NR_125364.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | NR_125364.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | NR_125364.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | NR_125364.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | NR_125364.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | NR_125364.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | NR_125364.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | NR_125364.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | NR_125364.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | NR_125364.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | NR_125364.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012633.hg.1 | NR_125364.1 | Refseq |

MKLN1-AS
Genomic Map