| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010018.hg.1 |
ENST00000534076.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010018.hg.1 |
ENST00000534076.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010018.hg.1 |
ENST00000534076.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010018.hg.1 |
ENST00000534076.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010018.hg.1 |
ENST00000534076.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010018.hg.1 |
NR_125366.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010018.hg.1 |
NR_125366.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010018.hg.1 |
NR_125366.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010018.hg.1 |
NR_125366.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010018.hg.1 |
NR_125366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100175714.hg.1 |
ENST00000534076.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100175714.hg.1 |
NR_125366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100175714.hg.1 |
ENST00000534076.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100175714.hg.1 |
NR_125366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100175714.hg.1 |
TC1100010018.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100010018.hg.1 |
ENST00000534076.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010018.hg.1 |
ENST00000534076.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010018.hg.1 |
ENST00000534076.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010018.hg.1 |
ENST00000534076.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010018.hg.1 |
ENST00000534076.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010018.hg.1 |
NR_125366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010018.hg.1 |
NR_125366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010018.hg.1 |
NR_125366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010018.hg.1 |
NR_125366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010018.hg.1 |
NR_125366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100175716.hg.1 |
ENST00000534076.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100175716.hg.1 |
NR_125366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100175716.hg.1 |
ENST00000534076.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100175716.hg.1 |
NR_125366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100175716.hg.1 |
TC1100010018.hg.1 |
Clariom D TC |