Assay Details
| Gene Symbol: LOC101927543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125419.1 | 2 - 3 | 205 | 104 | ||
| GenBank mRNA | AK057448.1 | - | 2 - 3 | 205 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MDM2 binding protein | MTBP | MDM2BP | -242.1kb | Sense | Chr. 8 - 120445398 - 120523635 on NCBI Build GRCh38 |
| Downstream | hyaluronan synthase 2 | HAS2 | 842.4kb | Anti-sense | Chr. 8 - 121613031 - 121641390 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101927543
Genomic Map