Assay Details
| Gene Symbol: CIB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125435.1 | 1 - 2 | 373 | 69 | ||
| GenBank mRNA | AI817087.1 | - | 1 - 2 | 109 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SH2 domain containing 7 | SH2D7 | -19.7kb | Anti-sense | Chr. 15 - 78089895 - 78104051 on NCBI Build GRCh38 | |
| Downstream | isocitrate dehydrogenase 3 (NAD(+)) alpha | IDH3A | 17.8kb | Anti-sense | Chr. 15 - 78149326 - 78170542 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Clariom D Assay | PSR1500175998.hg.1 | ENST00000258930.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175998.hg.1 | ENST00000539011.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175998.hg.1 | ENST00000557846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175998.hg.1 | NM_001271888.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175998.hg.1 | NM_001271889.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175998.hg.1 | NM_001301224.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175998.hg.1 | NM_006383.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175998.hg.1 | NR_125435.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175998.hg.1 | TC1500010134.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500175998.hg.1 | ENST00000258930.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175998.hg.1 | ENST00000539011.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175998.hg.1 | ENST00000557846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175998.hg.1 | NM_001271888.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175998.hg.1 | NM_001271889.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175998.hg.1 | NM_001301224.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175998.hg.1 | NM_006383.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175998.hg.1 | NR_125435.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | ENST00000258930.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | ENST00000539011.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | ENST00000557846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | ENST00000557917.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | ENST00000559054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | ENST00000559645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | ENST00000560618.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | ENST00000561190.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | NM_001271888.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | NM_001271889.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | NM_001301224.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | NM_006383.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | NR_125435.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | TC1500010134.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | ENST00000258930.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | ENST00000539011.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | ENST00000557846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | ENST00000557917.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | ENST00000559054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | ENST00000559645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | ENST00000560618.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | ENST00000561190.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | NM_001271888.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | NM_001271889.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | NM_001301224.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | NM_006383.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175993.hg.1 | NR_125435.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175999.hg.1 | ENST00000258930.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175999.hg.1 | ENST00000539011.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175999.hg.1 | NM_001271888.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175999.hg.1 | NM_001271889.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175999.hg.1 | NM_001301224.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175999.hg.1 | NM_006383.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175999.hg.1 | NR_125435.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175999.hg.1 | TC1500010134.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500175999.hg.1 | ENST00000258930.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175999.hg.1 | ENST00000539011.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175999.hg.1 | NM_001271888.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175999.hg.1 | NM_001271889.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175999.hg.1 | NM_001301224.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175999.hg.1 | NM_006383.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175999.hg.1 | NR_125435.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500176000.hg.1 | ENST00000258930.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500176000.hg.1 | ENST00000539011.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500176000.hg.1 | NM_001271888.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500176000.hg.1 | NM_001271889.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500176000.hg.1 | NM_001301224.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500176000.hg.1 | NM_006383.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500176000.hg.1 | NR_125435.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500176000.hg.1 | TC1500010134.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500176000.hg.1 | ENST00000258930.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500176000.hg.1 | ENST00000539011.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500176000.hg.1 | NM_001271888.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500176000.hg.1 | NM_001271889.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500176000.hg.1 | NM_001301224.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500176000.hg.1 | NM_006383.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500176000.hg.1 | NR_125435.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175996.hg.1 | ENST00000258930.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175996.hg.1 | ENST00000539011.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175996.hg.1 | ENST00000557846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175996.hg.1 | ENST00000557917.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175996.hg.1 | ENST00000559054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175996.hg.1 | ENST00000560618.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175996.hg.1 | NM_001271888.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175996.hg.1 | NM_001271889.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175996.hg.1 | NM_001301224.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175996.hg.1 | NM_006383.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175996.hg.1 | NR_125435.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175996.hg.1 | TC1500010134.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500175996.hg.1 | ENST00000258930.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175996.hg.1 | ENST00000539011.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175996.hg.1 | ENST00000557846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175996.hg.1 | ENST00000557917.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175996.hg.1 | ENST00000559054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175996.hg.1 | ENST00000560618.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175996.hg.1 | NM_001271888.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175996.hg.1 | NM_001271889.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175996.hg.1 | NM_001301224.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175996.hg.1 | NM_006383.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175996.hg.1 | NR_125435.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175992.hg.1 | ENST00000258930.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175992.hg.1 | ENST00000557917.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175992.hg.1 | ENST00000559054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175992.hg.1 | ENST00000559645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175992.hg.1 | ENST00000561190.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175992.hg.1 | NM_001301224.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175992.hg.1 | NM_006383.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175992.hg.1 | NR_125435.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175992.hg.1 | TC1500010134.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500175992.hg.1 | ENST00000258930.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175992.hg.1 | ENST00000557917.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175992.hg.1 | ENST00000559054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175992.hg.1 | ENST00000559645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175992.hg.1 | ENST00000561190.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500175992.hg.1 | NM_001301224.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175992.hg.1 | NM_006383.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500175992.hg.1 | NR_125435.1 | Refseq |

CIB2
Genomic Map