| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008961.hg.1 |
ENST00000428276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008961.hg.1 |
ENST00000428276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008961.hg.1 |
ENST00000428276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008961.hg.1 |
NR_125753.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008961.hg.1 |
NR_125753.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008961.hg.1 |
NR_125753.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300160479.hg.1 |
ENST00000428276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300160479.hg.1 |
NR_125753.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300160479.hg.1 |
ENST00000428276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300160479.hg.1 |
NR_125753.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300160479.hg.1 |
TC1300008961.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300008961.hg.1 |
ENST00000428276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008961.hg.1 |
ENST00000428276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008961.hg.1 |
ENST00000428276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008961.hg.1 |
NR_125753.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008961.hg.1 |
NR_125753.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008961.hg.1 |
NR_125753.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300160480.hg.1 |
ENST00000428276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300160480.hg.1 |
NR_125753.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300160480.hg.1 |
ENST00000428276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300160480.hg.1 |
NR_125753.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300160480.hg.1 |
TC1300008961.hg.1 |
Clariom D TC |