Assay Details
| Gene Symbol: LINC01048 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125762.1 | 3 - 4 | 267 | 107 | ||
| GenBank mRNA | BX091314.1 | - | 3 - 4 | 273 | 107 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | casein kinase 1 alpha 1 like | CSNK1A1L | CK1 | -375.8kb | Sense | Chr. 13 - 37103260 - 37105664 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 547 | LINC00547 | 50.5kb | Anti-sense | Chr. 13 - 37534940 - 37551536 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC01048
Genomic Map