| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015724.hg.1 |
ENST00000411433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015724.hg.1 |
ENST00000411433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015724.hg.1 |
ENST00000411433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015724.hg.1 |
ENST00000411433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015724.hg.1 |
NR_125777.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015724.hg.1 |
NR_125777.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015724.hg.1 |
NR_125777.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015724.hg.1 |
NR_125777.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015724.hg.1 |
NR_125777.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200215429.hg.1 |
NR_125777.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200215429.hg.1 |
NR_125777.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200215429.hg.1 |
TC0200015724.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200015724.hg.1 |
ENST00000411433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015724.hg.1 |
ENST00000411433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015724.hg.1 |
ENST00000411433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015724.hg.1 |
ENST00000411433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015724.hg.1 |
NR_125777.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015724.hg.1 |
NR_125777.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015724.hg.1 |
NR_125777.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015724.hg.1 |
NR_125777.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015724.hg.1 |
NR_125777.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200215430.hg.1 |
ENST00000411433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200215430.hg.1 |
NR_125777.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200215430.hg.1 |
ENST00000411433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200215430.hg.1 |
NR_125777.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200215430.hg.1 |
TC0200015724.hg.1 |
Clariom D TC |