Assay Details
| Gene Symbol: ZFPM2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125796.1 | 7 - 8 | 856 | 99 | ||
| NR_125797.1 | 4 - 5 | 408 | 99 | |||
| GenBank mRNA | BF244158.1 | - | 4 - 5 | 373 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | LDL receptor related protein 12 | LRP12 | ST7 | -1.2mb | Sense | Chr. 8 - 104489231 - 104589024 on NCBI Build GRCh38 |
| Downstream | oxidation resistance 1 | OXR1 |
Nbla00307, TLDC3 |
487.7kb | Anti-sense | Chr. 8 - 106270178 - 106752694 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000509144.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000509144.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000509144.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000509144.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000521622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000521622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000521622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000521622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125796.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125796.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125796.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125796.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125796.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125796.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125796.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125796.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125797.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125797.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125797.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125797.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011417.hg.1 | NR_125797.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177773.hg.1 | ENST00000509144.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177773.hg.1 | ENST00000520078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177773.hg.1 | ENST00000520078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177773.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177773.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177773.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177773.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177773.hg.1 | ENST00000521622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177773.hg.1 | NR_125797.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177773.hg.1 | ENST00000509144.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177773.hg.1 | ENST00000520078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177773.hg.1 | ENST00000520078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177773.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177773.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177773.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177773.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177773.hg.1 | ENST00000521622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177773.hg.1 | NR_125797.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177773.hg.1 | TC0800011417.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000509144.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000509144.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000509144.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000509144.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000518932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000520594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000521622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000521622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000521622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000521622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | ENST00000524045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125796.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125796.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125796.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125796.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125796.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125796.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125796.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125796.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125797.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125797.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125797.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125797.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011417.hg.1 | NR_125797.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177757.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177757.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177757.hg.1 | NR_125796.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177757.hg.1 | NR_125797.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177757.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177757.hg.1 | ENST00000520433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177757.hg.1 | NR_125796.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177757.hg.1 | NR_125797.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177757.hg.1 | TC0800011417.hg.1 | Clariom D TC |

ZFPM2-AS1
Genomic Map