| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008713.hg.1 |
ENST00000507332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008713.hg.1 |
ENST00000507332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008713.hg.1 |
ENST00000507933.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008713.hg.1 |
ENST00000507933.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008713.hg.1 |
ENST00000507933.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008713.hg.1 |
ENST00000514222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008713.hg.1 |
ENST00000514222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008713.hg.1 |
ENST00000514222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008713.hg.1 |
ENST00000514222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008713.hg.1 |
ENST00000514222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008713.hg.1 |
NR_125883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008713.hg.1 |
NR_125883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008713.hg.1 |
NR_125883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008713.hg.1 |
NR_125883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008713.hg.1 |
NR_125883.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008713.hg.1 |
NR_125884.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008713.hg.1 |
NR_125884.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008713.hg.1 |
NR_125884.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008713.hg.1 |
NR_125884.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008713.hg.1 |
ENST00000507332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008713.hg.1 |
ENST00000507332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008713.hg.1 |
ENST00000507933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008713.hg.1 |
ENST00000507933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008713.hg.1 |
ENST00000507933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008713.hg.1 |
ENST00000514222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008713.hg.1 |
ENST00000514222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008713.hg.1 |
ENST00000514222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008713.hg.1 |
ENST00000514222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008713.hg.1 |
ENST00000514222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008713.hg.1 |
NR_125883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008713.hg.1 |
NR_125883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008713.hg.1 |
NR_125883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008713.hg.1 |
NR_125883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008713.hg.1 |
NR_125883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008713.hg.1 |
NR_125884.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008713.hg.1 |
NR_125884.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008713.hg.1 |
NR_125884.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008713.hg.1 |
NR_125884.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400160054.hg.1 |
NR_125883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400160054.hg.1 |
NR_125884.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400160054.hg.1 |
NR_125883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400160054.hg.1 |
NR_125884.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400160054.hg.1 |
TC0400008713.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400160055.hg.1 |
NR_125883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400160055.hg.1 |
NR_125884.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400160055.hg.1 |
NR_125883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400160055.hg.1 |
NR_125884.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400160055.hg.1 |
TC0400008713.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400160061.hg.1 |
ENST00000507332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400160061.hg.1 |
ENST00000507933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400160061.hg.1 |
ENST00000514222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400160061.hg.1 |
NR_125883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400160061.hg.1 |
NR_125884.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400160061.hg.1 |
ENST00000507332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400160061.hg.1 |
ENST00000507933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400160061.hg.1 |
ENST00000514222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400160061.hg.1 |
NR_125883.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400160061.hg.1 |
NR_125884.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400160061.hg.1 |
TC0400008713.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400160068.hg.1 |
ENST00000507332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400160068.hg.1 |
NR_125884.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400160068.hg.1 |
ENST00000507332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400160068.hg.1 |
NR_125884.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400160068.hg.1 |
TC0400008713.hg.1 |
Clariom D TC |