Assay Details
| Gene Symbol: LOC102723766 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125933.1 | 1 - 2 | 164 | 181 | ||
| NR_125934.1 | 1 - 2 | 164 | 181 | |||
| GenBank mRNA | BG617470.1 | - | 1 - 2 | 162 | 181 | |
| BI835291.1 | - | 1 - 2 | 122 | 181 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC728175 | LOC728175 | -22.5kb | Sense | Chr. 4 - 184341031 - 184353977 on NCBI Build GRCh38 | |
| Downstream | interferon regulatory factor 2 | IRF2 | IRF-2 | 5.4kb | Sense | Chr. 4 - 184387722 - 184474572 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000512674.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000512674.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000512674.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000512674.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000512674.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000512674.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000512674.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000512674.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000512674.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000608785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000608785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000608785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000608785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000608785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000610223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000610223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000610223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000610223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000610223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000610223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000610223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000610683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000610683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000610683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000610683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000610683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000610683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000610683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000617323.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000617323.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000617323.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000617323.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000617323.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000617323.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000617323.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000617323.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | ENST00000617323.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012604.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186234.hg.1 | ENST00000610223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186234.hg.1 | ENST00000610683.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186234.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186234.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186234.hg.1 | ENST00000610223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186234.hg.1 | ENST00000610683.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186234.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186234.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186234.hg.1 | TC0400012604.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400186236.hg.1 | ENST00000608785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186236.hg.1 | ENST00000610223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186236.hg.1 | ENST00000610683.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186236.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186236.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186236.hg.1 | ENST00000608785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186236.hg.1 | ENST00000610223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186236.hg.1 | ENST00000610683.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186236.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186236.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186236.hg.1 | TC0400012604.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400186245.hg.1 | ENST00000608785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186245.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186245.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186245.hg.1 | ENST00000608785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186245.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186245.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186245.hg.1 | TC0400012604.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400186235.hg.1 | ENST00000608785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186235.hg.1 | ENST00000610223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186235.hg.1 | ENST00000610683.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186235.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186235.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186235.hg.1 | ENST00000608785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186235.hg.1 | ENST00000610223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186235.hg.1 | ENST00000610683.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186235.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186235.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186235.hg.1 | TC0400012604.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000512674.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000512674.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000512674.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000512674.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000512674.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000512674.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000512674.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000512674.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000512674.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000608785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000608785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000608785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000608785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000608785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000610223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000610223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000610223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000610223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000610223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000610223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000610223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000610683.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000610683.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000610683.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000610683.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000610683.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000610683.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000610683.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000617323.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000617323.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000617323.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000617323.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000617323.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000617323.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000617323.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000617323.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | ENST00000617323.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012604.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186244.hg.1 | ENST00000608785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186244.hg.1 | ENST00000610223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186244.hg.1 | ENST00000610683.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186244.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186244.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186244.hg.1 | ENST00000608785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186244.hg.1 | ENST00000610223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186244.hg.1 | ENST00000610683.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186244.hg.1 | NR_125933.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186244.hg.1 | NR_125934.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186244.hg.1 | TC0400012604.hg.1 | Clariom D TC |

LOC102723766
Genomic Map