Assay Details
| Gene Symbol: LOC101928977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125970.1 | 3 - 4 | 451 | 107 | ||
| GenBank mRNA | DA297063.1 | - | 3 - 4 | 452 | 107 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928995 | LOC101928995 | Anti-sense | Chr. 1 - 115919376 - 115925908 on NCBI Build GRCh38 | ||
| Downstream | solute carrier family 22 member 15 | SLC22A15 |
FLIPT1, PRO34686 |
51.5kb | Anti-sense | Chr. 1 - 115976498 - 116070054 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101928977
Genomic Map