| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006743.hg.1 |
ENST00000444276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006743.hg.1 |
ENST00000444276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006743.hg.1 |
ENST00000444276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006743.hg.1 |
ENST00000444276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006743.hg.1 |
ENST00000449895.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006743.hg.1 |
ENST00000449895.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006743.hg.1 |
ENST00000449895.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006743.hg.1 |
ENST00000449895.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006743.hg.1 |
ENST00000449895.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006743.hg.1 |
ENST00000449895.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006743.hg.1 |
ENST00000449895.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006743.hg.1 |
ENST00000449895.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006743.hg.1 |
NR_125999.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006743.hg.1 |
NR_125999.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006743.hg.1 |
NR_125999.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006743.hg.1 |
NR_125999.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006743.hg.1 |
NR_125999.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006743.hg.1 |
NR_125999.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006743.hg.1 |
ENST00000444276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006743.hg.1 |
ENST00000444276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006743.hg.1 |
ENST00000444276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006743.hg.1 |
ENST00000444276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006743.hg.1 |
ENST00000449895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006743.hg.1 |
ENST00000449895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006743.hg.1 |
ENST00000449895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006743.hg.1 |
ENST00000449895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006743.hg.1 |
ENST00000449895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006743.hg.1 |
ENST00000449895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006743.hg.1 |
ENST00000449895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006743.hg.1 |
ENST00000449895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006743.hg.1 |
NR_125999.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006743.hg.1 |
NR_125999.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006743.hg.1 |
NR_125999.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006743.hg.1 |
NR_125999.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006743.hg.1 |
NR_125999.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006743.hg.1 |
NR_125999.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146708.hg.1 |
ENST00000449895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146708.hg.1 |
ENST00000449895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146708.hg.1 |
NR_125999.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146708.hg.1 |
ENST00000449895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146708.hg.1 |
ENST00000449895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146708.hg.1 |
NR_125999.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146708.hg.1 |
TC0100006743.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100146710.hg.1 |
ENST00000444276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146710.hg.1 |
ENST00000449895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146710.hg.1 |
ENST00000449895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146710.hg.1 |
NR_125999.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146710.hg.1 |
ENST00000444276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146710.hg.1 |
ENST00000449895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146710.hg.1 |
ENST00000449895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146710.hg.1 |
NR_125999.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146710.hg.1 |
TC0100006743.hg.1 |
Clariom D TC |