Assay Details
| Gene Symbol: LOC102724552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125999.1 | 128 | ||||
| GenBank mRNA | BX114148.1 | - | 128 | |||
More Information
Updated Mapping Information
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100006743.hg.1 | ENST00000444276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006743.hg.1 | ENST00000444276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006743.hg.1 | ENST00000444276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006743.hg.1 | ENST00000444276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006743.hg.1 | ENST00000449895.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006743.hg.1 | ENST00000449895.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006743.hg.1 | ENST00000449895.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006743.hg.1 | ENST00000449895.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006743.hg.1 | ENST00000449895.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006743.hg.1 | ENST00000449895.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006743.hg.1 | ENST00000449895.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006743.hg.1 | ENST00000449895.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006743.hg.1 | NR_125999.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006743.hg.1 | NR_125999.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006743.hg.1 | NR_125999.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006743.hg.1 | NR_125999.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006743.hg.1 | NR_125999.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006743.hg.1 | NR_125999.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146715.hg.1 | ENST00000449895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146715.hg.1 | ENST00000449895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146715.hg.1 | NR_125999.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146715.hg.1 | ENST00000449895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146715.hg.1 | ENST00000449895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146715.hg.1 | NR_125999.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146715.hg.1 | TC0100006743.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100146716.hg.1 | NR_125999.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146716.hg.1 | NR_125999.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146716.hg.1 | TC0100006743.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100006743.hg.1 | ENST00000444276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006743.hg.1 | ENST00000444276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006743.hg.1 | ENST00000444276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006743.hg.1 | ENST00000444276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006743.hg.1 | ENST00000449895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006743.hg.1 | ENST00000449895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006743.hg.1 | ENST00000449895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006743.hg.1 | ENST00000449895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006743.hg.1 | ENST00000449895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006743.hg.1 | ENST00000449895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006743.hg.1 | ENST00000449895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006743.hg.1 | ENST00000449895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006743.hg.1 | NR_125999.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006743.hg.1 | NR_125999.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006743.hg.1 | NR_125999.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006743.hg.1 | NR_125999.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006743.hg.1 | NR_125999.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006743.hg.1 | NR_125999.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146712.hg.1 | ENST00000444276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146712.hg.1 | ENST00000449895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146712.hg.1 | ENST00000449895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146712.hg.1 | NR_125999.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146712.hg.1 | ENST00000444276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146712.hg.1 | ENST00000449895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146712.hg.1 | ENST00000449895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146712.hg.1 | NR_125999.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146712.hg.1 | TC0100006743.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100146717.hg.1 | NR_125999.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146717.hg.1 | NR_125999.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146717.hg.1 | TC0100006743.hg.1 | Clariom D TC |

LOC102724552
Genomic Map