| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017912.hg.1 |
ENST00000444721.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017912.hg.1 |
ENST00000444721.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017912.hg.1 |
NR_046582.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017912.hg.1 |
NR_046582.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017912.hg.1 |
ENST00000444721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017912.hg.1 |
ENST00000444721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017912.hg.1 |
NR_046582.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017912.hg.1 |
NR_046582.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100234995.hg.1 |
ENST00000444721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100234995.hg.1 |
NR_046582.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100234995.hg.1 |
ENST00000444721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100234995.hg.1 |
NR_046582.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100234995.hg.1 |
TC0100017912.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100234994.hg.1 |
ENST00000444721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100234994.hg.1 |
NR_046582.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100234994.hg.1 |
ENST00000444721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100234994.hg.1 |
NR_046582.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100234994.hg.1 |
TC0100017912.hg.1 |
Clariom D TC |