| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008586.hg.1 |
ENST00000510827.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008586.hg.1 |
ENST00000510827.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008586.hg.1 |
ENST00000510827.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008586.hg.1 |
ENST00000510827.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008586.hg.1 |
ENST00000510827.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008586.hg.1 |
ENST00000510827.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008586.hg.1 |
ENST00000515464.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008586.hg.1 |
ENST00000515464.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008586.hg.1 |
ENST00000515464.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008586.hg.1 |
NR_046629.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008586.hg.1 |
NR_046629.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008586.hg.1 |
NR_046629.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300161143.hg.1 |
ENST00000510827.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161143.hg.1 |
ENST00000510827.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161143.hg.1 |
NR_046629.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300161143.hg.1 |
ENST00000510827.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161143.hg.1 |
ENST00000510827.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161143.hg.1 |
NR_046629.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300161143.hg.1 |
TC0300008586.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300161145.hg.1 |
ENST00000510827.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161145.hg.1 |
ENST00000510827.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161145.hg.1 |
ENST00000515464.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161145.hg.1 |
NR_046629.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300161145.hg.1 |
ENST00000510827.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161145.hg.1 |
ENST00000510827.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161145.hg.1 |
ENST00000515464.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161145.hg.1 |
NR_046629.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300161145.hg.1 |
TC0300008586.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300008586.hg.1 |
ENST00000510827.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008586.hg.1 |
ENST00000510827.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008586.hg.1 |
ENST00000510827.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008586.hg.1 |
ENST00000510827.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008586.hg.1 |
ENST00000510827.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008586.hg.1 |
ENST00000510827.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008586.hg.1 |
ENST00000515464.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008586.hg.1 |
ENST00000515464.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008586.hg.1 |
ENST00000515464.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008586.hg.1 |
NR_046629.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008586.hg.1 |
NR_046629.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008586.hg.1 |
NR_046629.1 |
Refseq |