| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016926.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016926.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016926.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016926.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016926.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016926.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016926.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016926.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016926.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016926.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016926.hg.1 |
ENST00000421159.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016926.hg.1 |
ENST00000421159.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016926.hg.1 |
ENST00000421159.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016926.hg.1 |
ENST00000421449.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016926.hg.1 |
ENST00000421449.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016926.hg.1 |
ENST00000421449.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016926.hg.1 |
ENST00000421449.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016926.hg.1 |
NR_046696.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016926.hg.1 |
NR_046696.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016926.hg.1 |
NR_046696.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016926.hg.1 |
NR_046696.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016926.hg.1 |
NR_046696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100228219.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228219.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228219.hg.1 |
NR_046696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100228219.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228219.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228219.hg.1 |
NR_046696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100228219.hg.1 |
TC0100016926.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100228221.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228221.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228221.hg.1 |
ENST00000421449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228221.hg.1 |
NR_046696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100228221.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228221.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228221.hg.1 |
ENST00000421449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228221.hg.1 |
NR_046696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100228221.hg.1 |
TC0100016926.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100228225.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228225.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228225.hg.1 |
ENST00000421449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228225.hg.1 |
NR_046696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100228225.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228225.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228225.hg.1 |
ENST00000421449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228225.hg.1 |
NR_046696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100228225.hg.1 |
TC0100016926.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100228226.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228226.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228226.hg.1 |
NR_046696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100228226.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228226.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228226.hg.1 |
NR_046696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100228226.hg.1 |
TC0100016926.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100016926.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016926.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016926.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016926.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016926.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016926.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016926.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016926.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016926.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016926.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016926.hg.1 |
ENST00000421159.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016926.hg.1 |
ENST00000421159.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016926.hg.1 |
ENST00000421159.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016926.hg.1 |
ENST00000421449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016926.hg.1 |
ENST00000421449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016926.hg.1 |
ENST00000421449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016926.hg.1 |
ENST00000421449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016926.hg.1 |
NR_046696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016926.hg.1 |
NR_046696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016926.hg.1 |
NR_046696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016926.hg.1 |
NR_046696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016926.hg.1 |
NR_046696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100228224.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228224.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228224.hg.1 |
ENST00000421159.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228224.hg.1 |
ENST00000421449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228224.hg.1 |
NR_046696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100228224.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228224.hg.1 |
ENST00000413035.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228224.hg.1 |
ENST00000421159.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228224.hg.1 |
ENST00000421449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228224.hg.1 |
NR_046696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100228224.hg.1 |
TC0100016926.hg.1 |
Clariom D TC |