Assay Details
| Gene Symbol: NTM-IT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046820.1 | 2 - 3 | 372 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NTM antisense RNA 1 | NTM-AS1 | C11orf39 | -621.2kb | Anti-sense | Chr. 11 - 131662129 - 131663556 on NCBI Build GRCh38 |
| Downstream | opioid binding protein/cell adhesion molecule like | OPCML |
IGLON1, OBCAM, OPCM |
126.5kb | Anti-sense | Chr. 11 - 132403361 - 133532983 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

NTM-IT
Genomic Map