| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000425533.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000425533.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000425533.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000425533.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000425533.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000450093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000450093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000450093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000450093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000451672.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000451672.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000451672.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000451672.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000453396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000453396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000453396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000453396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000453396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000453396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000453396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000453396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000453396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000453396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000458036.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000458036.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000458036.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000458036.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000458036.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000540066.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000540066.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
ENST00000540066.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
NR_033991.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
NR_033991.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
NR_033991.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
NR_033991.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
NR_033991.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
NR_033992.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
NR_033992.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
NR_033992.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
NR_033992.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
NR_033992.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
NR_102376.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
NR_102376.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
NR_102376.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
NR_102376.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
NR_102376.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
NR_102377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
NR_102377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
NR_102377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006945.hg.1 |
NR_102377.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000425533.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000425533.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000425533.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000425533.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000425533.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000450093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000450093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000450093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000450093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000451672.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000451672.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000451672.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000451672.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000453396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000453396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000453396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000453396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000453396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000453396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000453396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000453396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000453396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000453396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000458036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000458036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000458036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000458036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000458036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000540066.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000540066.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
ENST00000540066.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
NR_033991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
NR_033991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
NR_033991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
NR_033991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
NR_033991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
NR_033992.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
NR_033992.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
NR_033992.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
NR_033992.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
NR_033992.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
NR_102376.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
NR_102376.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
NR_102376.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
NR_102376.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
NR_102376.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
NR_102377.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
NR_102377.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
NR_102377.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006945.hg.1 |
NR_102377.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900147139.hg.1 |
ENST00000425533.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900147139.hg.1 |
NR_102376.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900147139.hg.1 |
ENST00000425533.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900147139.hg.1 |
NR_102376.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900147139.hg.1 |
TC0900006945.hg.1 |
Clariom D TC |