| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007524.hg.1 |
ENST00000561916.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007524.hg.1 |
ENST00000561916.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007524.hg.1 |
ENST00000561916.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007524.hg.1 |
ENST00000561916.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007524.hg.1 |
NR_102400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007524.hg.1 |
NR_102400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007524.hg.1 |
NR_102400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007524.hg.1 |
NR_102400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
ENST00000247470.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
ENST00000247470.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
ENST00000247470.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
ENST00000247470.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
ENST00000247470.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
ENST00000247470.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
ENST00000247470.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
ENST00000247470.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
ENST00000350605.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
ENST00000350605.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
ENST00000350605.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
ENST00000350605.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
ENST00000350605.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
ENST00000561508.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
ENST00000561508.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
ENST00000561508.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
ENST00000561508.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
ENST00000561508.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
ENST00000561508.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
ENST00000561508.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
ENST00000565022.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
ENST00000565022.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
ENST00000565022.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
ENST00000565022.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
ENST00000565022.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
NM_013258.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
NM_013258.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
NM_013258.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
NM_013258.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
NM_013258.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
NM_013258.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
NM_145182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
NM_145182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
NM_145182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
NM_145182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010011.hg.1 |
NM_145182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007524.hg.1 |
ENST00000561916.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007524.hg.1 |
ENST00000561916.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007524.hg.1 |
ENST00000561916.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007524.hg.1 |
ENST00000561916.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007524.hg.1 |
NR_102400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007524.hg.1 |
NR_102400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007524.hg.1 |
NR_102400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007524.hg.1 |
NR_102400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600155133.hg.1 |
ENST00000561916.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600155133.hg.1 |
NR_102400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600155133.hg.1 |
TC1600007524.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600155133.hg.1 |
ENST00000561916.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600155133.hg.1 |
NR_102400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600155135.hg.1 |
ENST00000561916.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600155135.hg.1 |
NR_102400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600155135.hg.1 |
TC1600007524.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600155135.hg.1 |
ENST00000561916.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600155135.hg.1 |
NR_102400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
ENST00000247470.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
ENST00000247470.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
ENST00000247470.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
ENST00000247470.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
ENST00000247470.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
ENST00000247470.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
ENST00000247470.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
ENST00000247470.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
ENST00000350605.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
ENST00000350605.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
ENST00000350605.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
ENST00000350605.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
ENST00000350605.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
ENST00000561508.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
ENST00000561508.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
ENST00000561508.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
ENST00000561508.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
ENST00000561508.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
ENST00000561508.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
ENST00000561508.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
ENST00000565022.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
ENST00000565022.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
ENST00000565022.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
ENST00000565022.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
ENST00000565022.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
NM_013258.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
NM_013258.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
NM_013258.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
NM_013258.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
NM_013258.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
NM_013258.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
NM_145182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
NM_145182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
NM_145182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
NM_145182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010011.hg.1 |
NM_145182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600155134.hg.1 |
ENST00000561916.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600155134.hg.1 |
NR_102400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600155134.hg.1 |
TC1600007524.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600155134.hg.1 |
ENST00000561916.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600155134.hg.1 |
NR_102400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600155132.hg.1 |
ENST00000561916.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600155132.hg.1 |
NR_102400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600155132.hg.1 |
TC1600007524.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600155132.hg.1 |
ENST00000561916.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600155132.hg.1 |
NR_102400.1 |
Refseq |