| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011241.hg.1 |
ENST00000600652.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011241.hg.1 |
ENST00000600652.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011241.hg.1 |
ENST00000600652.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011241.hg.1 |
ENST00000600652.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011241.hg.1 |
ENST00000600652.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011241.hg.1 |
ENST00000600652.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011241.hg.1 |
ENST00000602810.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011241.hg.1 |
ENST00000602810.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011241.hg.1 |
ENST00000602810.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011241.hg.1 |
ENST00000602810.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011241.hg.1 |
NR_103455.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011241.hg.1 |
NR_103455.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011241.hg.1 |
NR_103455.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011241.hg.1 |
NR_103455.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011241.hg.1 |
NR_103455.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011241.hg.1 |
NR_103455.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011241.hg.1 |
NR_103456.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011241.hg.1 |
NR_103456.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011241.hg.1 |
NR_103456.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011241.hg.1 |
NR_103456.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011241.hg.1 |
NR_103456.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011241.hg.1 |
ENST00000600652.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011241.hg.1 |
ENST00000600652.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011241.hg.1 |
ENST00000600652.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011241.hg.1 |
ENST00000600652.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011241.hg.1 |
ENST00000600652.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011241.hg.1 |
ENST00000600652.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011241.hg.1 |
ENST00000602810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011241.hg.1 |
ENST00000602810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011241.hg.1 |
ENST00000602810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011241.hg.1 |
ENST00000602810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011241.hg.1 |
NR_103455.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011241.hg.1 |
NR_103455.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011241.hg.1 |
NR_103455.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011241.hg.1 |
NR_103455.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011241.hg.1 |
NR_103455.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011241.hg.1 |
NR_103455.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011241.hg.1 |
NR_103456.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011241.hg.1 |
NR_103456.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011241.hg.1 |
NR_103456.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011241.hg.1 |
NR_103456.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011241.hg.1 |
NR_103456.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600175773.hg.1 |
ENST00000600652.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600175773.hg.1 |
ENST00000602810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600175773.hg.1 |
NR_103455.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600175773.hg.1 |
NR_103456.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600175773.hg.1 |
ENST00000600652.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600175773.hg.1 |
ENST00000602810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600175773.hg.1 |
NR_103455.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600175773.hg.1 |
NR_103456.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600175773.hg.1 |
TC0600011241.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600175774.hg.1 |
ENST00000600652.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600175774.hg.1 |
ENST00000602810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600175774.hg.1 |
NR_103455.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600175774.hg.1 |
NR_103456.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600175774.hg.1 |
ENST00000600652.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600175774.hg.1 |
ENST00000602810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600175774.hg.1 |
NR_103455.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600175774.hg.1 |
NR_103456.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600175774.hg.1 |
TC0600011241.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600175775.hg.1 |
ENST00000600652.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600175775.hg.1 |
NR_103455.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600175775.hg.1 |
NR_103456.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600175775.hg.1 |
ENST00000600652.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600175775.hg.1 |
NR_103455.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600175775.hg.1 |
NR_103456.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600175775.hg.1 |
TC0600011241.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600175772.hg.1 |
ENST00000600652.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600175772.hg.1 |
NR_103455.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600175772.hg.1 |
ENST00000600652.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600175772.hg.1 |
NR_103455.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600175772.hg.1 |
TC0600011241.hg.1 |
Clariom D TC |