| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000413192.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000413192.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000413192.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000413192.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000413192.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000413192.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000413192.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000413192.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009980.hg.1 |
NR_103561.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100172385.hg.1 |
ENST00000413192.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100172385.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100172385.hg.1 |
NR_103561.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100172385.hg.1 |
ENST00000413192.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100172385.hg.1 |
ENST00000432386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100172385.hg.1 |
NR_103561.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100172385.hg.1 |
TC0100009980.hg.1 |
Clariom D TC |