| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012449.hg.1 |
ENST00000419364.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012449.hg.1 |
ENST00000419364.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012449.hg.1 |
ENST00000419364.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012449.hg.1 |
ENST00000419364.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012449.hg.1 |
ENST00000456467.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012449.hg.1 |
ENST00000456467.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012449.hg.1 |
ENST00000456467.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012449.hg.1 |
NR_103785.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012449.hg.1 |
NR_103785.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012449.hg.1 |
NR_103785.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012449.hg.1 |
NR_103785.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012449.hg.1 |
NR_103786.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012449.hg.1 |
NR_103786.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012449.hg.1 |
NR_103786.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012449.hg.1 |
NR_103786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012449.hg.1 |
ENST00000419364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012449.hg.1 |
ENST00000419364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012449.hg.1 |
ENST00000419364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012449.hg.1 |
ENST00000419364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012449.hg.1 |
ENST00000456467.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012449.hg.1 |
ENST00000456467.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012449.hg.1 |
ENST00000456467.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012449.hg.1 |
NR_103785.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012449.hg.1 |
NR_103785.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012449.hg.1 |
NR_103785.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012449.hg.1 |
NR_103785.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012449.hg.1 |
NR_103786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012449.hg.1 |
NR_103786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012449.hg.1 |
NR_103786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012449.hg.1 |
NR_103786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200190897.hg.1 |
ENST00000419364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200190897.hg.1 |
NR_103785.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200190897.hg.1 |
NR_103786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200190897.hg.1 |
ENST00000419364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200190897.hg.1 |
NR_103785.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200190897.hg.1 |
NR_103786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200190897.hg.1 |
TC0200012449.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200190898.hg.1 |
ENST00000419364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200190898.hg.1 |
ENST00000456467.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200190898.hg.1 |
NR_103785.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200190898.hg.1 |
NR_103786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200190898.hg.1 |
ENST00000419364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200190898.hg.1 |
ENST00000456467.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200190898.hg.1 |
NR_103785.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200190898.hg.1 |
NR_103786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200190898.hg.1 |
TC0200012449.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200190899.hg.1 |
ENST00000419364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200190899.hg.1 |
ENST00000456467.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200190899.hg.1 |
NR_103785.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200190899.hg.1 |
NR_103786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200190899.hg.1 |
ENST00000419364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200190899.hg.1 |
ENST00000456467.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200190899.hg.1 |
NR_103785.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200190899.hg.1 |
NR_103786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200190899.hg.1 |
TC0200012449.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200190904.hg.1 |
ENST00000419364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200190904.hg.1 |
NR_103786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200190904.hg.1 |
ENST00000419364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200190904.hg.1 |
NR_103786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200190904.hg.1 |
TC0200012449.hg.1 |
Clariom D TC |