| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011792.hg.1 |
ENST00000595010.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011792.hg.1 |
ENST00000595010.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011792.hg.1 |
ENST00000595010.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011792.hg.1 |
ENST00000600253.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011792.hg.1 |
ENST00000600253.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011792.hg.1 |
ENST00000600253.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011792.hg.1 |
ENST00000600253.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011792.hg.1 |
NR_103847.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011792.hg.1 |
NR_103847.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011792.hg.1 |
NR_103847.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011792.hg.1 |
NR_103847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900202431.hg.1 |
ENST00000595010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900202431.hg.1 |
ENST00000600253.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900202431.hg.1 |
NR_103847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900202431.hg.1 |
TC1900011792.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900202431.hg.1 |
NR_103847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900202431.hg.1 |
ENST00000595010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900202431.hg.1 |
ENST00000600253.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011792.hg.1 |
ENST00000595010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011792.hg.1 |
ENST00000595010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011792.hg.1 |
ENST00000595010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011792.hg.1 |
ENST00000600253.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011792.hg.1 |
ENST00000600253.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011792.hg.1 |
ENST00000600253.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011792.hg.1 |
ENST00000600253.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011792.hg.1 |
NR_103847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011792.hg.1 |
NR_103847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011792.hg.1 |
NR_103847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011792.hg.1 |
NR_103847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900202439.hg.1 |
ENST00000595010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900202439.hg.1 |
ENST00000600253.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900202439.hg.1 |
NR_103847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900202439.hg.1 |
TC1900011792.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900202439.hg.1 |
ENST00000595010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900202439.hg.1 |
ENST00000600253.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900202439.hg.1 |
NR_103847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900202430.hg.1 |
ENST00000600253.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900202430.hg.1 |
NR_103847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900202430.hg.1 |
TC1900011792.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900202430.hg.1 |
ENST00000600253.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900202430.hg.1 |
NR_103847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900202441.hg.1 |
ENST00000595010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900202441.hg.1 |
NR_103847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900202441.hg.1 |
TC1900011792.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900202441.hg.1 |
ENST00000595010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900202441.hg.1 |
NR_103847.1 |
Refseq |